Z171700
Ace double-jacketed coiled condenser
jacket L 400 mm
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.43
Produits recommandés
Joint
joint ST/NS24/40
Fabricant/nom de marque
Ace Glass 595515
L de l'enveloppe
400 mm
Description générale
Extra cooling capacity for volatile materials. Compact design retards diffusion.