Principaux documents
Z154253
Sigma-Aldrich
Aldrich® double-chamber vacuum trap
W × H 97 mm × 250 mm
About This Item
Code UNSPSC :
41104812
Nomenclature NACRES :
NB.43
Larg. × H
97 mm × 250 mm
Description générale
Inlet and outlet have serrated fittings, 14 to 16 mm. Can be used in a 2 L Dewar flask.
Informations légales
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC