Z152439
Aldrich® mercury bubbler
joint: ST/NS 24/40, capacity 40 mL
Synonyme(s) :
bubblers, scientific bubblers
About This Item
Produits recommandés
Joint
joint: ST/NS24/40
Capacité
40 mL
Description générale
Permits increased pressure within reaction vessels. Internal reservoir prevents mercury from being drawn into reaction flask or pushed out of bubbler. Do not overfill with mercury. Allows solvents to boil at higher temperatures.
Informations légales
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC