Z147958
Aldrich® reverse-flow bubbler
capacity 25 mL , per chamber
Synonyme(s) :
bubblers, scientific bubblers
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.43
Produits recommandés
Capacité
25 mL , per chamber
Description générale
Monitors gas flow into, or evolution from a system.
Informations légales
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC