Z107174
Lattice-rod clamp
die cast aluminum (alloy)
About This Item
Code UNSPSC :
41122413
Nomenclature NACRES :
NB.35
Produits recommandés
Matériaux
die cast aluminum (alloy)
Conditionnement
set of 12 ea
Description générale
Equipped with Allen screws. Assembles without dismantling the existing lattice-rod setup.
Application
For use with 1/2 in. diam. lattice rods.