Principaux documents
SYNS191530
Synthware metering valve stopcock plug
bore size 4 mm, Plug Size: 15.2/30
Synonyme(s) :
Synthware S191530
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.51
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware S191530
Taille de l'alésage
4 mm
Description générale
With a threaded PTFE needle valve for the fine control of flow through the bore of the stopcock plug. This replacement kit is complete with all the components.
Informations légales
Synthware is a trademark of Kemtech America Inc.