Principaux documents
SYNF221250
Synthware four neck round bottom flask with angled side necks
capacity 250 mL, center joint: ST/NS 14/20, side joint: ST/NS 14/20
Synonyme(s) :
Synthware F221250, 4-neck reaction flask
About This Item
Code UNSPSC :
41121804
Nomenclature NACRES :
NB.51
Matériaux
borosilicate glass flask
four-neck
round bottom
Joint
center joint: ST/NS14/20
side joint: ST/NS14/20
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware F221250
Capacité
250 mL
Description générale
Heavy wall design. All sizes are hand-blown from tubing to ensure uniform wall thickness.
Informations légales
Synthware is a trademark of Kemtech America Inc.