SYNC114200
Synthware™ cold finger condenser
joint: ST/NS 24/40, hose size 10 mm
Synonyme(s) :
Synthware C114200, Cold finger condenser with drip tip
About This Item
Produits recommandés
Joint
joint: ST/NS24/40
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware C114200
Taille du tuyau
10 mm
Description générale
Extended offset lower tip for drop control.
Informations légales
Synthware is a trademark of Kemtech America Inc.