SYNA351922
Synthware™ distillation adapter
top joint: ST/NS 19/22, bottom joint: ST/NS 19/22, side joint: 19/22, side arm angle: 75 deg
Synonyme(s) :
Synthware A351922, vacuum adapter, distillation adapter
About This Item
Produits recommandés
Joint
bottom joint: ST/NS19/22
side joint: 19/22
joint: ST/NS top19/22
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware A351922