Principaux documents
SYNA351014
Synthware distillation adapter
top joint: ST/NS 10/18, immersion depth 50 mm, side arm angle: 75 deg
Synonyme(s) :
Synthware A351014, adapter, sidearm adapter
About This Item
Code UNSPSC :
41121815
Nomenclature NACRES :
NB.51
Joint
joint: ST/NS top10/18
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware A351014
Profondeur d'immersion
50 mm
Description générale
For use as a component in atmospheric or vacuum distillations.
Informations légales
Synthware is a trademark of Kemtech America Inc.