SYNA282424
Synthware™ anti-splash adapter
100 mL, top joint: ST/NS 24/40, bottom joint: ST/NS 24/40
Synonyme(s) :
Synthware A282424, rotary evaporator bump trap, bump trap
About This Item
Produits recommandés
Joint
bottom joint: ST/NS24/40
joint: ST/NS top24/40
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware A282424
Capacité
100 mL