Principaux documents
MT51344821
Mettler-Toledo perfectION combination electrode
Measurement range 10-6-1 mol/L & 0.04-39000 mg/L
About This Item
Code UNSPSC :
39121436
Nomenclature NACRES :
NB.61
Matériaux
epoxy shaft
glass shaft (polymer)
Description
Lemo connector
Fabricant/nom de marque
Mettler-Toledo 51344821
Taille du câble
1.2 m
Description générale
Mettler Toledo perfectION ISE Combined ion-selective electrode means ISE measurement has never been so simple!
The new Click & Clear junction of the combined ion-selective electrodes combines three advantages in one tool: The electrolyte solution can flow easily through the junction by pressing the electrode head downwards. This ensures that the junction is optimally cleaned between the measurements and the electrolyte can be simply emptied and refilled if necessary.
Click & Clear is a sleeve junction that enables optimum contact between the electrolyte and measurement solution, even in difficult samples.
Features and Benefits
All combined ion-selective electrodes consist of a measuring electrode complete with an integrated reference. There is therefore no need for an additional reference electrode.
  • Sensor with attached cable and BNC connector
  • Reference electrolyte (60 mL)
  • Operating instructions
    To ensure an unproblematic start to your ISE measurement, the required calibration standards and ISA solutions (Ionic Strength Adjuster) can be ordered at the same time.
comb K Lemo
Informations légales
Click & Clear is a trademark of Mettler-Toledo, Inc.
perfectION is a trademark of Mettler-Toledo, Inc.