Principaux documents
BR704522
BRAND® discharge tube seripettor® /seripettor® pro
for 2 & 10 mL
Synonyme(s) :
Accessories, Bottle-top Dispenser, Dispenser, seripettor®
About This Item
Code UNSPSC :
41121500
Nomenclature NACRES :
NB.43
Matériaux
PTFE
Conditionnement
pack of 1 ea
Fabricant/nom de marque
BRAND 704522
Longueur
800 mm
Description générale
Tubing PTFE, coiled, length 800 mm, with safety handle. Not suited for peroxide.
Informations légales
BRAND is a registered trademark of BRAND GMBH + CO KG
seripettor is a registered trademark of BRAND GMBH + CO KG