Principaux documents
BR704350
BRAND® Bottle Thread Adapter
polypropylene, thread for GL 45 (Outer), for use with bottle thread S50
About This Item
Code UNSPSC :
41121516
Nomenclature NACRES :
SB.56
Matériaux
polypropylene
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
BRAND 704350
Raccords
thread for GL 45 (Outer)
Compatibilité
for use with bottle thread S50
Informations légales
BRAND is a registered trademark of BRAND GMBH + CO KG