pour consulter les tarifs organisationnels et contractuels.
Changer de vue
A propos de cet article
NACRES:
MA.24
UNSPSC Code:
41111761
Diam.:
3 mm
Packaging:
pack of 25 (per vial)
Service technique
Besoin d'aide ? Notre équipe de scientifiques expérimentés est là pour vous.
Laissez-nous vous aiderpackaging
pack of 25 (per vial)
diam.
3 mm
General description
Custom Mo 4-post lift-out grids for in-situ lift-out. Include multiple indexed mounting locations, two with vertical bars attachment surfaces and two with “V” shaped alignment surfaces. Sides have lower profile for easier access to outermost posts.
Still not finding the right product?
Explore all of our products under OMNI Probe Molybdenum 4-Post Lift-Out Grid