Merck
  • Dual Aurora A and JAK2 kinase blockade effectively suppresses malignant transformation.
Dual Aurora A and JAK2 kinase blockade effectively suppresses malignant transformation.
Oncotarget (2014-06-17)
Hua Yang, Harshani R Lawrence, Aslamuzzaman Kazi, Harsukh Gevariya, Ronil Patel, Yunting Luo, Uwe Rix, Ernst Schonbrunn, Nicholas J Lawrence, Said M Sebti
ABSTRACT
Aurora A and JAK2 kinases are involved in cell division and tumor cell survival, respectively. Here we demonstrate that ectopic expression of Aurora A and JAK2 together is more effective than each alone at inducing non-transformed cells to grow in an anchorage-independent manner and to invade. Furthermore, siRNA silencing or pharmacological inhibition of Aurora A and JAK2 with Alisertib and Ruxolitinib, respectively, is more effective than blocking each kinase alone at suppressing anchorage-dependent and -independent growth and invasion as well as at inducing apoptosis. Importantly, we have developed dual Aurora and JAK inhibitors, AJI-214 and AJI-100, which potently inhibit Aurora A, Aurora B and JAK2 in vitro. In human cancer cells, these dual inhibitors block the auto-phosphorylation of Aurora A (Thr-288) and the phosphorylation of the Aurora B substrate histone H3 (Ser-10) and the JAK2 substrate STAT3 (Tyr-705). Furthermore, AJI-214 and AJI-100 inhibit anchorage dependent and independent cell growth and invasion and induce G2/M cell cycle accumulation and apoptosis. Finally, AJI-100 caused regression of human tumor xenografts in mice. Taken together, our genetic and pharmacological studies indicate that targeting Aurora A and JAK2 together is a more effective approach than each kinase alone at inhibiting malignant transformation and warrant further advanced pre clinical investigations of dual Aurora A/JAK2 inhibitors as potential anti tumor agents.
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
Nocodazole, ≥99% (TLC), powder
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Aurka
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard