Merck
  • Three novel degraded steroids from cultures of the Basidiomycete Antrodiella albocinnamomea.
Three novel degraded steroids from cultures of the Basidiomycete Antrodiella albocinnamomea.
Steroids (2014-06-01)
Zi-Ming Chen, Xiao-Yan Yang, Qiong-Ying Fan, Zheng-Hui Li, Kun Wei, He-Ping Chen, Tao Feng, Ji-Kai Liu
ABSTRACT
Three novel degraded steroids, named albocisterols A-C (1-3), have been isolated from cultures of Antrodiella albocinnamomea. Their structures were defined by comprehensive spectroscopic analysis and single crystal X-ray crystallography. The mixture of compounds 2 and 3 exhibited significant inhibitory activities against protein tyrosine phosphatase 1B (PTP1B).
Sigma-Aldrich
Phosphatase substrate, powder
Sigma-Aldrich
Ursolic acid, ≥90%
Sigma-Aldrich
p-Nitrophenyl Phosphate Liquid Substrate System, liquid
Supelco
Ursolic acid, analytical standard
Ursolic acid, European Pharmacopoeia (EP) Reference Standard
Ursolic acid, primary reference standard