Merck
  • Indirect spectrophotometric determination of propranolol hydrochloride and piroxicam in pure and pharmaceutical formulations.
Indirect spectrophotometric determination of propranolol hydrochloride and piroxicam in pure and pharmaceutical formulations.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry (2002-06-27)
Babu G Gowda, Jaldappa Seetharamappa, Mahaveer B Melwanki
ABSTRACT
Two simple and sensitive indirect spectrophotometric methods for the assay of propranolol hydrochloride (PPH) and piroxicam (PX) in pure and pharmaceutical formulations have been proposed. The methods are based on the oxidation of PPH by a known excess of standard N-bromosuccinimide (NBS) and PX by ceric ammonium sulfate (CAS) in an acidic medium followed by the reaction of excess oxidant with promethazine hydrochloride (PMH) and methdilazine hydrochloride (MDH) to yield red-colored products. The absorbance values decreased linearly with increasing concentration of the drugs. The systems obeyed Beer's law over the concentration ranges of 0.5 - 12.5 and 0.3 - 16.0 microg/ml for PPH, and 0.4 - 7.5 and 0.2 - 10 microg/ml for PX with PMH and MDH, respectively. Molar absorptivity values, as calculated from Beer's law data, were found to be 1.36 x 10(4) and 2.55 x 10(4) l mol(-1) cm(-1) for PPH, and 2.08 x 10(4) and 2.05 x 10(4) l mol(-1) cm(-1) for PX with PMH and MDH, respectively. The common excipients and additives did not interfere with their determinations. The proposed methods have been successfully applied to the determinations of PPH and PX in various dosage forms. The results obtained by the proposed methods compare favorably with those of official methods.
Sigma-Aldrich
Ammonium cerium(IV) sulfate dihydrate
Sigma-Aldrich
Ammonium cerium(IV) sulfate dihydrate, ACS reagent, ≥94%
Sigma-Aldrich
Ammonium cerium(IV) sulfate dihydrate, puriss. p.a., ACS reagent, ≥98.0% (RT)