Tropine
Synonym(s):
3-Tropanol
Empirical Formula (Hill Notation):
C8H15NO
CAS Number:
Molecular Weight:
141.21
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Functional Group
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing