
Iron(III) chloride

Synonym(s):
Ferric chloride, Iron trichloride, Molysite, Iron(III) chloride
Linear Formula:
FeCl3
CAS Number:
Molecular Weight:
162.20
Products
Product Category
Brand
Sustainability
Feature
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Application
Reagent Type
Core
Special Grade
Technique
Showing 1-5 of 5
Compare
Product Number
Description
Pricing