CRM89078
Metazachlor Metabolite M479H004
certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
2-[(2,6-Dimethylphenyl)(1H-pyrazol-1-ylmethyl)amino]-2-oxoacetic acid, N-(2,6-Dimethylphenyl)-N-(1H-pyrazol-1-ylmethyl)oxalamide
About This Item
Recommended Products
grade
certified reference material
TraceCERT®
Quality Level
product line
TraceCERT®
form
solid
shelf life
limited shelf life, expiry date on the label
manufacturer/tradename
Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
SMILES string
[n]2(nccc2)CN(c1c(cccc1C)C)C(=O)C(=O)O
InChI key
PHMHHVKFXZNTKU-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com
Metazachlor metabolite M479H004 is a major degradation product of metazachlor. Metazachlor is a chloroacetamide herbicide, mainly used to control annual weeds and broadleaf weeds before or after germination. It is absorbed through the roots or stems and affects elongase activity during lipid biosynthesis, thereby inhibiting the synthesis of very long-chain fatty acids. Metazachlor also interferes with cell division and tissue differentiation during growth, resulting in the production of deformed seeds.
As per Regulation (EC) No 1107/2009, repealing the directive 91/414/ECC, metazachlor is approved for use in the European Union (EU). The maximum residue limit (MRL) for metazachlor presence in the crops varies between 0.02 mg/kg in most of the commodities to 0.9 mg/kg for turnip, Horse radishes, and Swedes/rutabagas in accordance with Regulation (EU) 2021/1807.
Application
The metazachlor metabolite M479H004 CRM may also find the following uses:
- To determine the effects of environmentally relevant concentrations of metazachlor and its metabolite on the marbled crayfish
- Determination of the metabolites of the herbicide metazachlor in agricultural crops by liquid chromatography-tandem mass spectrometry (LC-MS/MS)
Recommended products
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service