Key Documents
57356-U
Supelco
SPME Inlet Guide
greener alternative
About This Item
UNSPSC Code:
41115854
NACRES:
NB.21
Pricing and availability is not currently available.
material
aluminum
Quality Level
greener alternative product characteristics
Waste Prevention
Safer Solvents and Auxiliaries
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
technique(s)
solid phase microextraction (SPME): suitable
application(s)
food and beverages
greener alternative category
General description
We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Greener Chemistry. This is a Solid Phase Microextraction (SPME) technology product. SPME is one of the greener technologies for sample preparation as it is solvent free extraction technology and SPME fibres are re-usable and generates as little waste as possible, thus aligns with "Waste Prevention”, and ″Safer Solvents and Auxiliaries″. Click here for more information.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
Questions
  1. What GC instruments are suitable for use with the SPME inlet guide, 57356-U?
    1 answer
    1. The SPME inlet guide 57356-U is compatible with Varian systems with an auto-start, all HP's from the 5890 model onwards, and the newer models. It also fits the Merlin Microseal on the HP's. However, it is not designed to fit other models, and customizations for other models are not available.
      Helpful?