29305-U
Certified polypropylene caps with septa, pkg of 100
snap cap, PTFE/silicone septum
Synonym(s):
Certified Polypropylene Vial Caps with Seals
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41122001
NACRES:
NB.11
Recommended Products
material
PTFE/silicone septum
blue polypropylene cap
feature
closure type blue polypropylene snap cap
packaging
pkg of 100 ea
O.D. × H × I.D.
11.5 mm × 6 mm × 9 mm
fitting
thread for 11 mm
Looking for similar products? Visit Product Comparison Guide
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service