28432-U
24 mm/24 mm Polypropylene Viton® Connector
pk of 6
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
13111000
NACRES:
NB.21
Recommended Products
description
for connecting 24mm threaded glass hardware (Dioxin Columns & Top Flasks with 24 mm neck)
Quality Level
packaging
pk of 6
technique(s)
solid phase extraction (SPE): suitable
application(s)
food and beverages
Legal Information
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service