27519
Vials, screw top, graduated, open-top cap, preassembled
volume 15 mL, clear glass vial, thread for 18-400, O.D. × H 21 mm × 70 mm, PTFE/silicone septum
All Photos(1)
About This Item
UNSPSC Code:
41122001
Recommended Products
material
PTFE/silicone septum
black phenolic cap
clear glass
clear glass vial
feature
closure type black phenolic hole cap
closure type screw top vial
packaging
pkg of 100 ea
O.D. × H
21 mm × 70 mm
volume
15 mL
fitting
thread for 18-400
Related Categories
General description
Hole cap is black phenolic.
Septum is blue PTFE on white silicone.
Septum is blue PTFE on white silicone.
related product
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)