27037
Micro-Reaction Vessel, black phenolic solid cap, pkg of 12
volume 2 mL, clear glass vial, styrene-butadiene septum, O.D. × H 16 mm × 56 mm
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41122001
Recommended Products
material
clear glass vial
styrene-butadiene septum
feature
closure type black phenolic solid top cap
closure type screw top vial
graduations
pre-assembled
packaging
pkg of 12 ea
O.D. × H
16 mm × 56 mm
volume
2 mL
fitting
thread for 15-425
Looking for similar products? Visit Product Comparison Guide
Features and Benefits
- Micro Reaction Vessel of volume 2 mL is cone shaped for easy removal of small samples.
- Constructed of heavy wall borosilicate glass.
- Supplied with black phenolic solid top cap.
- Can be autoclaved or centrifuged.
related product
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Geraldine Dowling et al.
Drug testing and analysis (2018-03-16)
Nitrazolam and clonazolam are 2 designer benzodiazepines available from Internet retailers. There is growing evidence suggesting that such compounds have the potential to cause severe adverse events. Information about tolerability in humans is scarce but typically, low doses can be
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service