Skip to Content
Merck

22084-U

Swagelok® Connector to Male NPT

stainless steel, 1/8 in. Swagelok, 1/8 in. male NPT, 200-1-2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41112500
eCl@ss:
32035302
NACRES:
SB.54

Pricing and availability is not currently available.

material

stainless steel

packaging

pkg of 1 ea

manufacturer/tradename

200-1-2

fitting

1/8 in. Swagelok
1/8 in. male NPT

Looking for similar products? Visit Product Comparison Guide

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
237213S2252799416
assay

99.999% trace metals basis

assay

≥97.0%

assay

≥99.0%

assay

≥97%

impurities

≤15.0 ppm Trace Metal Analysis

impurities

≤0.01% insolubles

impurities

-

impurities

≤0.01% insolubles

mp

271 °C (lit.)

mp

271 °C (lit.)

mp

271 °C (lit.)

mp

271 °C (lit.)

autoignition temp.

914 °F

autoignition temp.

914 °F

autoignition temp.

914 °F

autoignition temp.

914 °F

General description

Nuts and ferrules included.

Legal Information

Swagelok is a registered trademark of Swagelok Company

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service