Key Documents
Z743535
Richter Anti-Vibration pads
L × W 7.0 in. × 8.5 in., pkg of 2
About This Item
UNSPSC Code:
41121800
NACRES:
NB.35
Pricing and availability is not currently available.
packaging
pkg of 2
manufacturer/tradename
Diversified Biotech VIBE-1000
L × W
7.0 in. × 8.5 in.
General description
Inexpensive solution which dampens vibrations generated from lab equipment
Creates a stable surface to keep equipment in place
Fits vortexes, shakers, rockers and microscopes
Small, lightweight design
Stack pads for heavier units or position side-by-side forlarger equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)