Skip to Content
Merck
All Photos(1)

Key Documents

Z688541

Angled bottle rest

accommodates 50 mL to 1 liter square or round bottles

Synonym(s):

angled bottle suport

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122800
NACRES:
NB.35

Pricing and availability is not currently available.

packaging

pack of 1 ea

manufacturer/tradename

Diversified Biotech REST-1000

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
Z742257Z742259Z742258
packaging

pack of 1 ea

packaging

pack of 1 ea

packaging

pack of 1 ea

packaging

pack of 1 ea

General description

This angled bottle rest is convenient for tilting the bottle to access material inside easier.

  • Comfort - Pipette at a comfortable 45°
  • Durable - Made from long-lasting acrylic and polypropylene
  • Ideally sized - Accommodates 50 mL to 1 liter square or round bottles
  • Space saving - Compact design takes up little benchtop space

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service