T2918
Nalgene® centrifuge tubes, Oak Ridge Style 3139
nominal capacity 50 mL
Synonym(s):
High-Speed Centrifuge Tubes
All Photos(1)
About This Item
UNSPSC Code:
41121703
NACRES:
NB.11
Recommended Products
material
cap (Sealing cap)
polypropylene
polypropylene copolymer
sterility
non-sterile
feature
RCF 100,605 × g
packaging
pack of 10 ea
manufacturer/tradename
Nalgene® 3139-0050
nominal capacity
50 mL
General description
Spin with confidence with when you use these Nalgene® Oak Ridge high-speed PPCO centrifuge tubes complete with sealing caps. These tubes are chemically compatible with a wider variety of laboratory reagents than polycarbonate centrifuge tubes and are repeately autoclavable for extended performance.
Features and Benefits
- Greater chemical resistance than polycarbonate
- Autoclavable
- Can be used up to 50,000 x g in refrigerated or non-refrigerated centrifuges
- 3139-0050 (T2918) can be used up to 100,605 x g in the T29-8x50 rotor
- Comes with polypropylene sealing caps with silicone gaskets for extra leak protection when spinning >10,000 x g or spinning hazardous materials
- Must be filled to 80% of total capacity when centrifuging
- Autoclavable/Leakproof† when used with supplied Nalgene closure; Repeated autoclaving will reduce the life of PC centrifuge tubes.
Packaging
With sealing cap
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)