SYNS371004
Synthware™ microliter syringe
borosilicate glass 3.3, needle, Bevel Tip, capacity 10 μL
Synonym(s):
Synthware S371004, glass sample syringe
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
Recommended Products
material
borosilicate glass 3.3
feature
needle Bevel Tip
packaging
pack of 1 ea
manufacturer/tradename
Kemtech S371004
capacity
10 μL
Looking for similar products? Visit Product Comparison Guide
General description
Microliter Sample Syringe, Capacity: 10ul
Application
kemetch; synthware; syringe; microliter
Features and Benefits
Organic Chemistry
Legal Information
Synthware is a trademark of Kemtech America Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service