SML3342
Trofinetide Trifluoroacetate
≥95% (HPLC)
Synonym(s):
Gly-2–methylPro–Glu Trifluoroacetate, Gly-2MePro-Glu Trifluoroacetate, Glycyl-2-methyl-L-prolyl-L-Glutamic acid Trifluoroacetate, NNZ 2566 Trifluoroacetate, NNZ-2566 Trifluoroacetate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Biochem/physiol Actions
Trofinetide is a more enzymatically stable analogue of the N-terminal tripeptide of IGF-1(GPE) that exhibits potent neuroprotective effects in varies animal models of neurodegenerative diseases and traumatic brain injury. Trofinetide treatment provides clinically meaningful improvement in Rett syndrome (RTT) and Fragile X syndrome (FXS).
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
NNZ-2566: a Gly-Pro-Glu analogue with neuroprotective efficacy in a rat model of acute focal stroke.
Journal of the neurological sciences, 278(1-2), 85-90 (2009-01-23)
The N-terminal cleavage product of human insulin-like growth factor-1 (IGF-1) in the brain is the tripeptide molecule Glypromate (Gly-Pro-Glu). Glypromate has demonstrated neuroprotective effects in numerous in vitro and in vivo models of brain injury and is in clinical trials
Pediatric neurology, 110, 30-41 (2020-07-15)
We analyze the safety and tolerability of trofinetide and provide a preliminary evaluation of its efficacy in adolescent and adult males with fragile X syndrome. This study was an exploratory, phase 2, multicenter, double-blind, placebo-controlled, parallel group study of the
Neuromolecular medicine, 15(3), 504-514 (2013-06-15)
The tripeptide glycine-proline-glutamate analogue NNZ-2566 (Neuren Pharmaceuticals) demonstrates neuroprotective efficacy in models of traumatic brain injury. In penetrating ballistic-like brain injury (PBBI), it significantly decreases injury-induced upregulation of inflammatory cytokines including TNF-α, IFN-γ, and IL-6. However, the mechanism by which
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service