SML1624
JNJ-63533054
≥98% (HPLC)
Synonym(s):
(S)-3-Chloro-N-(2-oxo-2-((1-phenylethyl)amino)ethyl)benzamide
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C17H17ClN2O2
CAS Number:
Molecular Weight:
316.78
UNSPSC Code:
12352200
NACRES:
NA.77
Recommended Products
Quality Level
Assay
≥98% (HPLC)
form
powder
optical activity
[α]/D -89 to -99°, c = 1 in ethanol
color
white to beige
solubility
DMSO: 20 mg/mL, clear
storage temp.
room temp
Biochem/physiol Actions
JNJ-63533054 is a potent and selective agonist of the orphan receptor hGPR139, a GPCR receptor expressed in the brain in circumventricular regions of the habenula and septum. JNJ-63533054 has an EC50 value of 16 nM, crosses the blood-brain barrier, and is orally available.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service