Skip to Content
Merck
All Photos(2)

Key Documents

SAB4502548

Sigma-Aldrich

Anti-BAX antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Apoptosis regulator BAX, BAX, BCL2L4, Bcl-2-like protein 4, Bcl2-L-4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 21 kDa

species reactivity

human, rat, mouse

concentration

~1 mg/mL

technique(s)

ELISA: 1:10000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... BAX(581)

General description

Anti-BAX Antibody detects endogenous levels of total BAX protein.

Immunogen

The antiserum was produced against synthesized peptide derived from human Bax.

Immunogen Range: 1-50

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qiong Yu et al.
The journal of obstetrics and gynaecology research, 47(11), 3789-3796 (2021-09-11)
To investigate the optimal dose of mifepristone and lithospermum combination regimen on medical abortion in early pregnancy rats without increasing side effects. Sixty sexually mature female Sprague Dawley (SD) rats with early pregnancy were randomly allocated into 10 groups, including
Yuan Wei et al.
Acta pharmaceutica (Zagreb, Croatia), 74(1), 131-147 (2024-03-30)
There is increasing evidence that long non-coding RNAs (lncRNAs) play a crucial role in the development and progression of malignant tumors, particularly pancreatic cancer. In this study, the influence of the lncRNA TINCR on the behavior of human pancreatic cancer

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service