Skip to Content
Merck
All Photos(1)

Key Documents

PZ0250

Sigma-Aldrich

PF-04859989 hydrochloride

≥98% (HPLC)

Synonym(s):

(3S)-3-Amino-1-hydroxy-3,4-dihydroquinolin-2(1H)-one; L-3-Amino-3,4-dihydro-1-hydroxycarbstyril hydrochloride, PF-4859989 hydrochloride, PF04859989 hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H10N2O2 · HCl
CAS Number:
Molecular Weight:
214.65
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

H2O: 20 mg/mL, clear

storage temp.

room temp

SMILES string

N[C@H]1CC2=CC=CC=C2N(O)C1=O.Cl

InChI

1S/C9H10N2O2.ClH/c10-7-5-6-3-1-2-4-8(6)11(13)9(7)12;/h1-4,7,13H,5,10H2;1H/t7-;/m0./s1

InChI key

WZOBDOKCHIUXAY-FJXQXJEOSA-N

Biochem/physiol Actions

PF-04859989 is a a brain-penetrable irreversible inhibitor of kynurenine amino transferase II (KAT II).
PF-04859989 is a brain-penetrable irreversible inhibitor of kynurenine amino transferase II (KAT II), the enzyme responsible for most of the brain synthesis of kynurenic acid, which has been implicated in several psychiatric and neurological disorders, including schizophrenia and bipolar disorder, and thought to impair cognitive function. PF-04859989 has IC50 values of 23 nM for hKAT II and 263 nM for rKAT II. PF-04859989 is ~1000-fold selective for KAT II over human KAT I, KAT III, and KAT IV. PF-04859989 prevented ketamine-induced disruption of performance in memory tasks in both rodents and nonhuman primates.

Features and Benefits

This compound is a featured product for Neuroscience research. Click here to discover more featured Neuroscience products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service