Skip to Content
Merck
All Photos(1)

Key Documents

MTCP70051S

MTC Bio ASPIR-8 Resevoir

capacity 5 mL, sterile, pack of 100 ea (individually wrapped)

Synonym(s):

cooled tube rack, reagent dish, reagent holder

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121522
NACRES:
NB.15

sterility

sterile

packaging

pack of 100 ea (individually wrapped)

manufacturer/tradename

MTC Bio P7005-1S

capacity

5 mL

Looking for similar products? Visit Product Comparison Guide

General description

ASPIR-8 Reservoir, 5mL, sterile, Individually wrapped, 100/cs

Features and Benefits

  • Graduated inner walls
  • Micro-trough U-shaped bottoms
  • Pouring spouts on all 4 corners

Legal Information

MTC is a trademark of MTC Bio

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service