MAK557
Melanin Assay Kit
sufficient for 100 fluorometric tests
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116104
NACRES:
NA.84
Recommended Products
detection method
fluorometric
relevant disease(s)
dermatological diseases; immunological diseases; neurological disorders
shipped in
wet ice
storage temp.
−20°C
Related Categories
General description
Melanin, a biological pigment found in various biological systems, exhibits a range of physiological properties. It is widely acknowledged for its ability to impact overall health through photoprotective and immunological actions. Additionally, melanin has antioxidant, anti-inflammatory, immunomodulatory, radioprotective, hepatic, gastrointestinal, neurological, and hypoglycemic effects. The Melanin Assay Kit uses a substrate that generates a fluorescent product upon reaction with melanins. The fluorescence intensity is proportional to the amount of melanin in a sample.
Suitability
The Melanin Assay Kit provides a simple and effective method to measure melanin content in cells and other biological samples.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Met. Corr. 1 - Ox. Liq. 3 - Skin Corr. 1B
Storage Class Code
5.1B - Oxidizing hazardous materials
WGK
WGK 2
Flash Point(F)
188.6 °F - closed cup
Flash Point(C)
87 °C - closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service