MAK451
Glucose-6-Phosphate Dehydrogenase Assay Kit
Sufficient for 100 colorimetric tests
Synonym(s):
G6PD Assay Kit
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12161503
NACRES:
NA.84
Recommended Products
Quality Level
input
plasma
serum
tissue
cell culture medium
detection method
colorimetric
relevant disease(s)
hematological disorder, anemia
storage temp.
−20°C
Related Categories
General description
Glucose-6-Phosphate Dehydrogenase (G6PDH) is a cytosolic enzyme in the pentose phosphate pathway which supplies reducing energy to cells by maintaining the level of the co-enzyme nicotinamide adenine dinucleotide phosphate (NADPH). G6PDH reduces nicotinamide adenine dinucleotide phosphate (NADP) to NADPH while oxidizing glucose-6-phosphate (G6P). Humans with a genetic deficiency of G6PDH are predisposed to non-immune hemolytic anemia.
Application
The Glucose-6-Phosphate Dehydrogenase Assay Kit may be used for:
- Metabolism Research
- Drug Discovery
Features and Benefits
- Efficient and Precise: Requires only 20 μL of sample and a short 15-minute reaction time while detecting glucose-6-phosphate dehydrogenase levels across a wide range from 0.2 to 100 U/L, offering both convenience and sensitivity.
- Compatibility with High-Throughput Systems: Effortlessly incorporate our kit into high-throughput handling systems, guaranteeing seamless integration and maintaining efficiency and accuracy in your laboratory operations.
Suitability
The kit is suitable for the quantitative determination of Glucose-6-Phosphate concentration and evaluation of drug effects on its metabolism in plasma, serum, tissue and culture media, etc.
Principle
The non-radioactive, colorimetric Glucose-6-Phosphate Dehydrogenase Assay Kit is based on the reduction of tetrazolium salt MTT in a NADPH-coupled enzymatic reaction to a reduced form of MTT which exhibits an absorption maximum at A565 nm. The increase in absorbance at A565 nm is proportional to the enzyme activity.
Other Notes
For additional information on our range of Biochemicals, please complete this form.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Muta. 2
Storage Class Code
12 - Non Combustible Liquids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service