Skip to Content
Merck
All Photos(1)

Key Documents

JN0001

Sigma-Aldrich

Alniditan dihydrochloride

≥98% (HPLC)

Synonym(s):

JNJ 16384810-AAC, N-[[(2R)-chroman-2-yl]methyl]-N′-(1,4,5,6-tetrahydropyrimidin-2-yl)propane-1,3-diamine dihydrochloride, R 91274, R091274

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H26N4O·2HCl
CAS Number:
Molecular Weight:
375.34
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D -65 to -75°, c = 1.0 in methanol

color

white to beige

solubility

H2O: 2 mg/mL, clear

storage temp.

room temp

SMILES string

[Cl-].N1CCCN=C1NCCCNC[C@@H]2Oc3c(cccc3)CC2.[H+]

InChI

1S/C17H26N4O.ClH/c1-2-6-16-14(5-1)7-8-15(22-16)13-18-9-3-10-19-17-20-11-4-12-21-17;/h1-2,5-6,15,18H,3-4,7-13H2,(H2,19,20,21);1H/t15-;/m1./s1

InChI key

BHFNHKKGGQTOBO-XFULWGLBSA-N

Biochem/physiol Actions

Alniditan is a potent and selective serotonin 5-HT1B/5-HT1D receptor agonist that exhibits antimigraine effects.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pankaj Bhalla et al.
European journal of pharmacology, 436(1-2), 23-33 (2002-02-09)
Using a combination of reverse transcription polymerase chain reaction (RT-PCR) and inverse-PCR techniques, we amplified, cloned and sequenced a full-length porcine 5-hydroxytryptamine 1F (5-ht(1F)) receptor complementary DNA (cDNA) derived from porcine trigeminal ganglion. Sequence analysis revealed 1101 base pairs (bp)
A S Lesage et al.
British journal of pharmacology, 123(8), 1655-1665 (1998-05-30)
1. Alniditan, a novel migraine abortive agent, is a potent 5-HT1B/5-HT1D receptor agonist of nM affinity. We compared the agonistic properties of alniditan, sumatriptan and dihydroergotamine on the cloned human 5-HT1B receptor expressed at 200 fmol mg(-1) protein (Bmax) in
J E Leysen et al.
Molecular pharmacology, 50(6), 1567-1580 (1996-12-01)
Alniditan is a new migraine-abortive agent. It is a benzopyran derivative and therefore structurally unrelated to sumatriptan and other indole-derivatives and to ergoline derivatives. The action of sumatriptan is thought to be mediated by 5-hydroxytryptamine (5-HT)1D-type receptors. We investigated the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service