HS2860H
TrueNorth® Cardboard Cryovial Box Cell Divider
Holds 81 x 1.5-2.0 mL tubes, pk of 12
Synonym(s):
cryovial storage, freezer box divider
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41123304
NACRES:
NB.22
Recommended Products
material
cardboard
feature
Holds 81 x 1.5-2.0 mL tubes
packaging
pk of 12
manufacturer/tradename
Heathrow Scientific HS2860H
technique(s)
cryopreservation: suitable
well diam.
12 mm
Looking for similar products? Visit Product Comparison Guide
Application
TrueNorth® Cardboard Cryovial Box Cell Divider may be suitable for the storage of small samples and tubes.
Features and Benefits
- Store in mechanical freezers or vapor phase LN2, -196ºC.
- Colors facilitate identifying and sorting purposes with ease.
- Bottom vent holes aid in faster circulation and elimination of cold air, vapor, or LN2
- All box footprints are 13.3 * 13.3 cm, select from two different heights
- Use tubes capable of being stored at the required temperature of the user.
- Not recommended for autoclaving.
Legal Information
TrueNorth is a registered trademark of Heathrow Scientific LLC
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service