Skip to Content
Merck
All Photos(1)

Key Documents

GE17-5448-01

Capto Blue

Cytiva 17-5448-01, pack of 25 mL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151817
NACRES:
NA.56

ligand

Cibacron Blue

packaging

pack of 25 mL

manufacturer/tradename

Cytiva 17-5448-01

storage condition

(Solution of 0.1 M KH2PO4 and 20% Ethanol; 0.1 M Potassium Phosphate containing 20% Ethanol)

matrix

highly cross-linked agarose

avg. part. size

75 μm (d50v)

cleaning in place

2-13

working range

3-13

capacity

>24 mg binding capacity(HSA/ml medium (Qb10 at 4 min residence time))

suitability

suitable for bioprocess medium

storage temp.

2-8°C

Related Categories

General description

Capto Blue affinity resin with Cibacron Blue ligand has outstanding pressure and flow properties, allowing rapid processing of large sample volumes of albumin and other proteins.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

Capto is a trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

This page shows how to convert between flow velocity and volumetric flow rate in affinity chromatography of antibodies.

Protocols

This page shows how purify or remove albumin with Blue Sepharose and Capto Blue from Cytiva.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service