Skip to Content
Merck
All Photos(1)

Key Documents

GE10600080

Amersham Protran® Premium Western blotting membranes, nitrocellulose

pore size 0.2 μm, sheet W × L 100 mm × 100 mm, pkg of 25 ea

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105339
NACRES:
NB.22

packaging

pkg of 25 ea

manufacturer/tradename

Cytiva 10600080

sheet W × L

100 mm × 100 mm

pore size

0.2 μm pore size

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Nitrocellulose blotting membrane, provides excellent sensitivity, resolution, and low background in fluorescence Western blotting detection applications.

Application

Amersham Protran® Premium Western blotting membranes, nitrocellulose may be used in:
  • Western blotting
  • Southern blotting
  • Northern blotting

Features and Benefits

  • Low auto-fluorescence across a wide range of excitation/emission wavelengths makes the membrane suitable for fluorescence-based immunodetection and multiplexing.
  • High binding capacity (173 – 203 µg IgG/cm2) of small proteins (< Mr 20 000) reduces risk of "blow-through".
  • Available in popular precut sizes for laboratory application.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Legal Information

Amersham is a trademark of Cytiva
Protran is a registered trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Precautionary Statements

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service