EP3125000150
Eppendorf® Research® Plus Move It pipette
no. of channels, 4, volume 30-300 μL, cone distance 9-33 mm, orange
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
Recommended Products
material
orange
feature
no. of channels 4
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 3125000150
cone distance
9-33 mm
volume
30-300 μL
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
- Adjustable tip spacing pipette
- Multi-channel pipette
- 4-channel
- 30-300μL
- 9mm cone distance
- Orange
Application
- Forward pipetting
- Reverse pipetting
- Removal of supernatants
- Sample mixing
- Phase extraction
- Filling of plates, gels, and reaction vessel
Features and Benefits
Air cushioned pipettes with variable volume and adjustable tip spacing
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Eppendorf Research is a registered trademark of Eppendorf AG
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service