EP0030089502
Eppendorf® ViscoTip, 10 mL, Eppendorf Quality™
pack of 100 ea (4 bags of 25 tips)
All Photos(1)
About This Item
UNSPSC Code:
41121600
NACRES:
NB.02
maximum volume:
10 mL
volume:
(10 mL)
sterility:
non-sterile
packaging:
pack of 100 ea (4 bags of 25 tips)
manufacturer/tradename:
Eppendorf® 0030089502
Recommended Products
material
orange
sterility
non-sterile
packaging
pack of 100 ea (4 bags of 25 tips)
manufacturer/tradename
Eppendorf® 0030089502
maximum volume
10 mL
volume
(10 mL)
Related Categories
Features and Benefits
- Tips are specialized for handling highly viscous liquids that have a dynamic viscosity from 200 mPa*s to 14,000 mPa*s
- Ideal for dispensing volumes ranging between 100 µL to 10 mL, with 10 µL increments
- New tip design increases ergonomics, and working speed and reduces energy consumption
- Save time on calculations with the Multipette® M4/E3/E3x, which automatically recognizes tips and calculates volume.
- Free of leachable such as slip agents, biocides, and plasticizers that interfere with experimental results
- ViscoTips come with a distinct color-coded double ring, and continuous volume scale, making it easier to differentiate them from Combitips advanced®
Legal Information
Combitips advanced is a registered trademark of Eppendorf AG
Eppendorf is a registered trademark of Eppendorf AG
Eppendorf Quality is a trademark of Eppendorf AG
Multipette is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)