Skip to Content
Merck
All Photos(1)

Key Documents

EHU114821

Sigma-Aldrich

MISSION® esiRNA

targeting human TERF1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CCCTTGATGCACAGTTTGAAAATGATGAACGAATTACACCCTTGGAATCAGCCCTGATGATTTGGGGTTCAATTGAAAAGGAACATGACAAACTTCATGAAGAAATACAGAATTTAATTAAAATTCAGGCTATAGCTGTTTGTATGGAAAATGGCAACTTTAAAGAAGCAGAAGAAGTCTTTGAAAGAATATTTGGTGATCCAAATTCTCATATGCCTTTCAAAAGCAAATTGCTTATGATAATCTCTCAGAAAGATACATTTCATTCCTTTTTTCAACACTTCAGCTACAACCACATGATGGAGAAAATTAAGAGTTATGTGAATTATGTGCTAAGTGAAAAATCATCAACCTTTCTAATGAAGGCAGCGGCAAAAGTAGTAGAAAGCAAAAGGACAAGAACAA

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Surya Shin Ichi Sutanto et al.
Journal of cell communication and signaling, 13(4), 523-530 (2019-06-17)
People with diabetes mellitus have shorter telomeres compared with non-diabetic subjects. The aim of this study was to investigate an in-vitro model of telomere shortening under diabetes metabolic conditions. The mechanisms of the accelerated telomere length attrition and the potential
Rosa Maria Porreca et al.
eLife, 9 (2020-01-15)
Telomeres are a significant challenge to DNA replication and are prone to replication stress and telomere fragility. The shelterin component TRF1 facilitates telomere replication but the molecular mechanism remains uncertain. By interrogating the proteomic composition of telomeres, we show that
Luxi Sun et al.
Nucleic acids research, 45(7), 3844-3859 (2017-02-06)
Werner syndrome (WS) is a progeroid-like syndrome caused by WRN gene mutations. WS cells exhibit shorter telomere length compared to normal cells, but it is not fully understood how WRN deficiency leads directly to telomere dysfunction. By generating localized telomere-specific
Lu Yang et al.
Nucleic acids research, 45(7), 3906-3921 (2017-02-06)
Oxidative DNA damage triggers telomere erosion and cellular senescence. However, how repair is initiated at telomeres is largely unknown. Here, we found unlike PARP1-mediated Poly-ADP-Ribosylation (PARylation) at genomic damage sites, PARylation at telomeres is mainly dependent on tankyrase1 (TNKS1). TNKS1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service