Skip to Content
Merck
All Photos(1)

Key Documents

EHU050781

Sigma-Aldrich

MISSION® esiRNA

targeting human LIN28B

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GCCCCTTGGATATTCCAGTCGATGTATTTGTACACCAAAGCAAACTATTCATGGAAGGATTTAGAAGCCTAAAAGAAGGAGAACCAGTGGAATTCACATTTAAAAAATCTTCCAAAGGCCTTGAGTCAATACGGGTAACAGGACCTGGTGGGAGCCCCTGTTTAGGAAGTGAAAGAAGACCCAAAGGGAAGACACTACAGAAAAGAAAACCAAAGGGAGATAGATGCTACAACTGTGGTGGCCTTGATCATCATGCTAAGGAATGTAGTCTACCTCCTCAGCCAAAGAAGTGCCATTACTGTCAGAGCATCATGCACATGGTGGCAAACTGCCCACATAAAAATGTTGCACAGCCACCCGCGAGTTCTCAGGGAAGACAGGAAGCAGAATCCCAGCCATGCACTTCAACTCTCCCTCGAGAAGTGGGAGGCGGGCATGGCTGTACATCACCAC

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jing Wu et al.
Acta biochimica et biophysica Sinica, 51(5), 455-462 (2019-04-09)
Choriocarcinoma is a rare and malignant trophoblastic tumor. However, the molecular mechanisms by which choriocarcinoma is regulated remain unknown. In the present study, we first elucidated that LIN28B was highly expressed in human choriocarcinoma tissues and choriocarcinoma cell lines. Our
Jianbiao Zhou et al.
Journal of hematology & oncology, 10(1), 138-138 (2017-07-12)
Current conventional chemotherapy for acute myeloid leukemia (AML) can achieve remission in over 70% of patients, but a majority of them will relapse within 5 years despite continued treatment. The relapse is postulated to be due to leukemia stem cells (LSCs)
Chong Chen et al.
Cancer research, 75(8), 1725-1735 (2015-03-07)
Considerable evidence suggests that proinflammatory pathways drive self-renewal of cancer stem-like cells (CSC), but the underlying mechanisms remain mainly undefined. Here we report that the let7 repressor LIN28B and its regulator IKBKB (IKKβ) sustain cancer cell stemness by interacting with
B-X Yan et al.
Oncogene, 33(45), 5288-5294 (2013-11-05)
Tumor drug resistance remains a major challenge in the treatment of cancer. Here, we show that Prostatic secretory protein 94 (PSP94) levels are reduced in ovarian cancer patients with high levels of excision repair cross-complementing 1 (ERCC1), a marker for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service