Skip to Content
Merck
All Photos(1)

Key Documents

EHU041711

Sigma-Aldrich

MISSION® esiRNA

targeting human CPT1A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CCACTGAGTCATGCGACTTCGTGCGGGCCATGGTGGACCCGGCCCAGACGGTGGAACAGAGGCTGAAGTTGTTCAAGTTGGCGTCTGAGAAGCATCAGCATATGTATCGCCTCGCCATGACCGGCTCTGGGATCGATCGTCACCTCTTCTGCCTTTACGTGGTGTCTAAATATCTCGCTGTGGAGTCCCCTTTCCTTAAGGAAGTTTTATCTGAGCCTTGGAGATTATCAACAAGCCAGACCCCTCAGCAGCAAGTGGAGCTGTTTGACTTGGAGAATAACCCAGAGTACGTGTCCAGCGGAGGGGGCTTTGGACCGGTTGCTGATGACGGCTATGGTGTGTCGTACATCCTTGTGGGAGAGAACCTCATCAATTTCCACATTTCTTCCAAGTTCTCTTGCCCTGAG

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Seher Balaban et al.
Molecular cancer research : MCR, 17(4), 949-962 (2019-01-17)
Prostate cancer cells exhibit altered cellular metabolism but, notably, not the hallmarks of Warburg metabolism. Prostate cancer cells exhibit increased de novo synthesis of fatty acids (FA); however, little is known about how extracellular FAs, such as those in the
Hideki Iwamoto et al.
Cell metabolism, 28(1), 104-117 (2018-06-05)
Intrinsic and evasive antiangiogenic drug (AAD) resistance is frequently developed in cancer patients, and molecular mechanisms underlying AAD resistance remain largely unknown. Here we describe AAD-triggered, lipid-dependent metabolic reprogramming as an alternative mechanism of AAD resistance. Unexpectedly, tumor angiogenesis in adipose
Jin Seok et al.
Stem cell research & therapy, 11(1), 1-1 (2020-01-05)
Human placenta-derived mesenchymal stem cells (PD-MSCs) are powerful sources for cell therapy in regenerative medicine. However, a limited lifespan by senescence through mechanisms that are well unknown is the greatest obstacle. In the present study, we first demonstrated the characterization
Trang Thi Thu Nguyen et al.
The Journal of clinical investigation, 130(7), 3699-3716 (2020-04-22)
The Warburg effect is a tumor-related phenomenon that could potentially be targeted therapeutically. Here, we showed that glioblastoma (GBM) cultures and patients' tumors harbored super-enhancers in several genes related to the Warburg effect. By conducting a transcriptome analysis followed by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service