CLS4117
Corning® DeckWorks™ standard pipet tips
1 - 200μL pipet tips, graduated, hinged racks, yellow, non-sterile, polypropylene, 96 tips/rack, 10 racks/pack, 4 packs/case
Synonym(s):
pipet tips, pipette tips
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121600
NACRES:
NB.01
volume range:
1-200 μL
sterility:
non-sterile
feature:
DNase free
RNase free
barrier: no
disposable
endotoxin free
filter: no
graduated
Pipet tip: round
RNase free
barrier: no
disposable
endotoxin free
filter: no
graduated
Pipet tip: round
packaging:
rack of 96
manufacturer/tradename:
Corning 4117
Recommended Products
material
polypropylene
yellow tip
sterility
non-sterile
feature
DNase free
RNase free
barrier: no
disposable
endotoxin free
filter: no
graduated
Pipet tip: round
packaging
rack of 96
manufacturer/tradename
Corning 4117
volume range
1-200 μL
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Corning® DeckWorks™ Pipet Tips
Bulk Pipet Tips
Corning DeckWorks bulk pipet tips are compatible with DeckWorks racks and packaged in resealable polyethylene bags. Pipet tip sizes are as follows: 10 uL, 200 uL and 1000 uL.
Universal Fit Pipet Tips
Corning universal fit tips in hinged DeckWorks racks are available in 10 uL, 200 uL, 300 uL and 1000 uL sizes. The 10 uL, 300 uL and 1000 uL tips are extended length and allow for improved access into micro (1.5 mL) and standard conical bottom (15/50 mL) centrifuge tubes.
Reload Systems
Robust 96-tip racks are designed for repeated steam autoclave cycles and can be reused by removing the empty grey tip deck and replacing it with a new reload deck of 96 tips from the DeckWorks reload system. Corning racked 300 uL DeckWorks pipet tips are designed for all popular 300 uL multi-channel instruments.
This system is designed to allow you to maximize storage space and help reduce the production and waste of plastic materials. Both the hinged racks and reload decks within this system are manufactured with recycled plastic. The pipet tips are made in a controlled molding environment to ensure accuracy and precision across all tip volumes.
Medical grade resins and production excellence allow for full certification as:
Bulk Pipet Tips
Corning DeckWorks bulk pipet tips are compatible with DeckWorks racks and packaged in resealable polyethylene bags. Pipet tip sizes are as follows: 10 uL, 200 uL and 1000 uL.
Universal Fit Pipet Tips
Corning universal fit tips in hinged DeckWorks racks are available in 10 uL, 200 uL, 300 uL and 1000 uL sizes. The 10 uL, 300 uL and 1000 uL tips are extended length and allow for improved access into micro (1.5 mL) and standard conical bottom (15/50 mL) centrifuge tubes.
Reload Systems
Robust 96-tip racks are designed for repeated steam autoclave cycles and can be reused by removing the empty grey tip deck and replacing it with a new reload deck of 96 tips from the DeckWorks reload system. Corning racked 300 uL DeckWorks pipet tips are designed for all popular 300 uL multi-channel instruments.
This system is designed to allow you to maximize storage space and help reduce the production and waste of plastic materials. Both the hinged racks and reload decks within this system are manufactured with recycled plastic. The pipet tips are made in a controlled molding environment to ensure accuracy and precision across all tip volumes.
Medical grade resins and production excellence allow for full certification as:
- Nonpyrogenic
- Human DNA-free
- RNase-/DNase-free
Legal Information
Corning is a registered trademark of Corning, Inc.
DeckWorks is a trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service