CLS3893
Corning® 1536 well plates
flat bottom clear polystyrene wells, TC-treated, 50/cs, sterile, lid
Synonym(s):
1536 multiwell plate, 1536 well microplate, 1536 well microtiter plate, 1536 well plate, HTS plate, TC-treated plate, immunoassay plate, tissue-culture treated plate
About This Item
flat bottom clear polystyrene wells
polystyrene plate
material
black plate
flat bottom clear polystyrene wells
polystyrene plate
sterility
sterile
feature
lid
skirt
packaging
pack of 10
case of 50
manufacturer/tradename
Corning 3893
maximum volume
12.5 μL
size
1536 wells
surface area
0.03 cm2
working volume
8 μL
color
clear
suitability
suitable for (fluorescent; luminescent assays)
binding type
Tissue Culture (TC)-treated surface
Looking for similar products? Visit Product Comparison Guide
General description
- 1536 well polystyrene plates
- Low background fluorescence, minimal light scatter and reduced crosstalk
- Excellent signal dynamic range due to low background fluorescence and enhanced signal intensity
- Opaque walls to prevent well-to-well crosstalk
- Flat well bottoms possess superior flatness -minimizes variability within a plate and between plates
- Round well bottoms for reduced air entrapment and improved CV′s and Z factor
- Raised well bottoms for high sensitivity
- Flood reservior on four sides to reduce instrument contamination
- Suitable for fluorescent and luminescent assays
- Designed for fluorescent homogeneous cell-based assays
- Compatible with standard readers and automation
- Compatible for bar coding
- Superior performance compared to competitor plates: Lower CVs, higher signal-to-noise ratios, and Lower background fluorescence
- Processed for superior flatness with minimal variability within plate and between plates
- Recommended working volume of up to 10μL
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service