Skip to Content
Merck
All Photos(1)

Key Documents

CLS3694

Corning® 96 Well Half-Area Microplate

flat bottom, black polystyrene, bag of 25, non-sterile, lid: no

Synonym(s):

cell culture plate, multi well plates, multiple well plates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.13
material:
black polystyrene
flat bottom
size:
96 wells
Pricing and availability is not currently available.

material

black polystyrene
flat bottom

sterility

non-sterile

feature

lid: no
skirt

packaging

bag of 25

manufacturer/tradename

Corning 3694

maximum volume

190 μL

size

96 wells

well D

10.7 mm

well working volume

25-125 μL

working volume

125 μL

color

black

suitability

suitable for (cell cuture)

binding type

non-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

Unique half area plates can help save on rare and expensive reagents by reducing the amount of reagent needed per well, while still being readable by most standard plate readers.

Features and Benefits

  • Corning® 96 Well Half-Area Microplate, flat bottom, black polystyrene, no lid has medium binding polystyrene surface
  • Favors minimal well-to-well crosstalk during fluorescent assays
  • Hydrophobic and binds biomolecules through passive interactions
  • Displays low background fluorescence and minimal light scattering
  • Immobilization of large molecules
  • Standard microplate footprint and dimensions

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service